Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Swingsette®, M515

Tissue processing/embedding cassettes, Swingsette®, M515

Orange
Catalog # 720-1629
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Swingsette®, M515
Catalog # 720-1629
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Swingsette®, M515
Tissue processing/embedding cassettes, Swingsette®, M515

Orange

Price
SFr. 306.00
Log in for availability
1 * 1.500 ST
Product Details & Documents

Acetal polymer.

  • Integral, hinged lid, with large tab for easy opening
  • Disposable plastic cassettes highly resistant to histological solvents
  • Flow-through slots maximise fluid exchange
  • Front face inclined 45° to enable use with leading automatic labelling systems.

 : Each case contains three boxes of 500 cassettes.

Specifications
  • Colour:
    Orange
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.