Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bossheads, three angles

Bossheads, three angles

Duraluminium, 19 - 23 mm
Catalog # BOCH5656
Supplier:   BOCHEM
Bossheads, three angles
Catalog # BOCH5656
Supplier:  BOCHEM
Bossheads, three angles
Bossheads, three angles

Duraluminium, 19 - 23 mm

Price
SFr. 32.80
Log in for availability
1 * 1 ST
Product Details & Documents

Three angled bossheads made of duraluminium (aluminium alloy).

  • Angle: 90 + 180 + 270°
  • Suitable for multilevel and three dimension constructions
Specifications
  • Material:
    Duraluminium
  • For rod Ø:
    19 - 23 mm