Orders LinkContactUsLinkComponent
VWR® MultiTray®/MultiBox® plus, Pipette Tips
VWR® MultiTray®/MultiBox® plus, Pipette Tips
10 µl tips, crystal, graduated, 0,1 - 10 µl, Transparent, MultiTray® 10×96 tips
Catalog # 612-2913
Supplier:   VWR Collection
VWR® MultiTray®/MultiBox® plus, Pipette Tips
Catalog # 612-2913
Supplier:  VWR Collection
VWR® MultiTray®/MultiBox® plus, Pipette Tips
VWR® MultiTray®/MultiBox® plus, Pipette Tips
10 µl tips, crystal, graduated, 0,1 - 10 µl, Transparent, MultiTray® 10×96 tips
Price
SFr. 91.30
Log in for availability
1 * 960 ST
Product Details & Documents
Tips racked in MultiTrays®. Compatible with most micropipettes, except 612-2913 which is compatible with Gilson® and Labmate® micropipettes. MultiBox® plus is an autoclavable PP box with dustproof, hinged and sliding lid, which may be opened completely or partially. MultiBox® plus is suitable for use with all MultiTrays®.
  • Tips fit firmly on micropipettor cones, have straight shape for optimum handling, and offer minimum fluid retention
  • MultiBox® plus can be repeatedly autoclaved, but should be placed in an adapted container to avoid contact with the autoclave walls (sterilisation conditions: +121 °C, 1,2 bar, max. 30 minutes, with vessels open)
  • Up to a maximum of five MultiBox® plus can be stacked
Ordering information: Tips supplied pre-assembled in MultiTray® systems.
Specifications
  • Packed:
    MultiTray® 10×96 tips
  • Volume:
    0,1 - 10 µl
  • Seq:
    02
  • Tip style:
    10 µl tips, crystal, graduated
  • Tip colour:
    Transparent