Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tripod stands

Tripod stands

Retort stand base, tripod, 1,7 kg, 185 mm
Catalog # 241-0341
Supplier:   USBECK Laborgeräte
Tripod stands
Catalog # 241-0341
Tripod stands
Tripod stands

Retort stand base, tripod, 1,7 kg, 185 mm

Price
56.10 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

Steel, powder coated.

  • With M10 threaded hole
  • No support rod
  • Non-slip, rubber feet ensure firm footing
Specifications
  • Description:
    Retort stand base, tripod
  • Arm length:
    185 mm
  • Weight:
    1,7 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.