VWR®, Calibration Weights, Class 1
5000 g, 12000 ± mg
Catalog # 611-8765
Supplier: VWR Collection
Jump to:
Product Details & Documents
Stainless steel masses adjusted to class 1 for precise checking and adjustment of electronic balances.
Certifications: ASTM certified.
Delivery information: Individual weights are supplied without certificate.
Documents
VWR® for Calibrations VWR® for Weighing
Product Literature
Specifications
- Weight:5000 g
- Seq:07
- Tolerance:12000 ± mg