Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Humidity standards

Humidity standards

Certified humidity standards, ±0,5% RH, 35%, 5 Ampoul
Catalog # 620-2224
Humidity standards
Catalog # 620-2224
CAS Number:  
Humidity standards
Humidity standards

Certified humidity standards, ±0,5% RH, 35%, 5 Ampoul

CAS Number:  
Price
91.20 SFr.
Log in for availability
1 * 5 Ampull
Product Details & Documents

Although most relative humidity (RH) probes have excellent long-term stability, it is recommended that their calibration be checked regularly.

  • Traceable to national standard – SCS (Swiss Calibration Service) certificate included
  • Ampoules contain unsaturated salt solutions (~0,8 ml)
  • Inexpensive on-site calibration
  • Simple and safe to use
  • 10 year shelf life
  • Practical packs of five ampoules of the same humidity value

 : Each ampoule is labelled with the humidity value and serial number.

 : Values and accuracy (measurement uncertainty values) below are measured at 23 °C.

Specifications

Specifications

  • Pk:
    5 Ampoul
  • Description:
    Certified humidity standards
  • Relative humidity (RH%):
    35%
  • Accuracy:
    ±0,5% RH

Specifications