Cyclohexanone ≥99.0% analytical reagent
25 L
Supplier: VWR Chemicals
Cyclohexanone ≥99.0% analytical reagent
Catalog # 23236.460
Supplier: VWR Chemicals
CAS Number: 108-94-1
Jump to:
Product Details & Documents
Synonyms: Pimelic Ketone
- CAS Number:108-94-1
- Formula:C₆H₁₀(=O)
- MDL Number:MFCD00001625
- Molecular Weight:98.14 g/mol
- Merck Index:13,02755
- Boiling Pt:155.6 °C (1013 hPa)
- Melting Pt:–31 °C
- Density:0.9478 g/cm³ (20 °C)
- Flash Pt:43 °C
- Storage Temperature:Ambient
- EINECS:203-631-1
- UN:1915
- ADR:3,III
- REACH:01-2119453616-35
Specifications
Specification Test Results
- Assay:Min. 99.0 %
- Acidity:Max. 0.005 meq/g
- Boiling point:154.5 - 156.5 °C
- Density (20/4):0.946 - 0.948
- Refractive index (20°C):1.449 - 1.451
- Viscosity (25°C):1.92 - 2.2 mPa·s
- Cyclohexanol:Max. 0.5 %
- Water:Max. 0.2 %
Specifications
- Pk:25 L
- Cat. No.:23236.460
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



