ContactUsLinkComponent
Semi-automatic, electro mechanically controlled vertical laboratory autoclave with UK-plug, Chamberdimension (xL) 380 x 690 mm Volu me: 85 liters 1 * 1 items
Catalog # 481-2071
Supplier:  TUTTNAUER
Semi-automatic, electro mechanically controlled vertical laboratory autoclave with UK-plug, Chamberdimension (xL) 380 x 690 mm Volu me: 85 liters 1 * 1 items
Catalog # 481-2071
Supplier:  TUTTNAUER
Frequently Bought Together