Tip refill system, Axygen®
Empty refill racks, RFL-R, 10 µl and 200 µl refill inserts
Catalog # 732-0958
Supplier: Corning
Jump to:
Product Details & Documents
The Axygen® Tip Refill System contains durable refill racks that can be easily refilled by hand and are autoclavable.
- RNase-/DNase-free
- Non pyrogenic
- Translucent rack allows tips to be easily seen
- Colour-coded insert allows for easy identification of tip
Specifications
- Description:Empty refill racks, RFL-R
- Seq:01
- For use with:10 µl and 200 µl refill inserts
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



