Orders LinkContactUsLinkComponent
Quartz control plates
Quartz control plates
Standard quartz control plate −30 °Z, −10 °A, 1
Catalog # 635-0405
Supplier:   BELLINGHAM STANLEY
Quartz control plates
Catalog # 635-0405
Supplier:  BELLINGHAM STANLEY
CAS Number:  
Quartz control plates
Quartz control plates
Standard quartz control plate −30 °Z, −10 °A, 1
CAS Number:  
Price
SFr. 1’790.00
Log in for availability
1 * 1 ST
Product Details & Documents
Quartz control plates for the calibration and verification of polarimeters. of optical and digital polarimeters and saccharimeters.
  • Inert quartz with relatively low temperature coefficient, easy to handle, and infinite shelf life
  • Available in three optical rotation values
  • Three decimal place uncertainty
The optional temperature saddles may used as an interface between the quartz plate and instrument temperature sensor to provide fine compensation.
Certifications: Traceable to PTB. UKAS-MRA calibration in accordance with ISO 17025:2005 between 325-880nm available.
Ordering information: Sadlles can be ordered in option.
Delivery information: Supplied with a certificate of verification in a plastic storage case.
Specifications
Specifications
  • Pk:
    1
  • Description:
    Standard quartz control plate −30 °Z, −10 °A