Orders LinkContactUsLinkComponent
Reference electrodes
Reference electrodes
Reference electrode, Ag/AgCl reference system, B 1491 HD, +5...+135 °C, Glass, KCI 3 mol/l, Ceramic, 12×170 mm, Screw plug head
Catalog # SCOI285111504
Supplier:   SI Analytics
Reference electrodes
Catalog # SCOI285111504
Supplier:  SI Analytics
Reference electrodes
Reference electrodes
Reference electrode, Ag/AgCl reference system, B 1491 HD, +5...+135 °C, Glass, KCI 3 mol/l, Ceramic, 12×170 mm, Screw plug head
Price
351.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
These reference electrodes are available with low maintenance plug heads or with fixed cables and free end.
Specifications
  • Connection:
    Screw plug head
  • Ø×L:
    12×170 mm
  • Type:
    B 1491 HD
  • Description:
    Reference electrode, Ag/AgCl reference system
  • Material:
    Glass
  • Diaphr.:
    Ceramic
  • Electrolyte:
    KCI 3 mol/l
  • Temp.:
    +5...+135 °C