Orders LinkContactUsLinkComponent
Extraction thimbles
27×80 mm
  537-0003
 :   PRAT DUMAS
Extraction thimbles
  537-0003
Extraction thimbles
Extraction thimbles
27×80 mm
SFr. 121.00
1 * 25 ST
Pure cotton cellulose
  • Neutral and free of fats and grease
  • Moulded in one piece
  • Very resistant to solvents
Applications include extraction of sludges, asphalt, foodstuffs and in the analysis and quality control of powders and dusts.
  • Int.Ø×H:
    27×80 mm