Extraction thimbles
30×60 mm
Catalog # 537-0004
Supplier: PRAT DUMAS
Jump to:
Product Details & Documents
Pure cotton cellulose
- Neutral and free of fats and grease
- Moulded in one piece
- Very resistant to solvents
Applications include extraction of sludges, asphalt, foodstuffs and in the analysis and quality control of powders and dusts.
Specifications
- Int.Ø×H:30×60 mm