Orders LinkContactUsLinkComponent
Extraction thimbles | Avantor
Extraction thimbles
58×170 mm
Catalog # 537-0012
Supplier:   PRAT DUMAS
Extraction thimbles
Catalog # 537-0012
Supplier:  PRAT DUMAS
Extraction thimbles
Extraction thimbles
58×170 mm
Price
SFr. 489.00
Log in for availability
1 * 25 ST
Product Details & Documents
Pure cotton cellulose
  • Neutral and free of fats and grease
  • Moulded in one piece
  • Very resistant to solvents
Applications include extraction of sludges, asphalt, foodstuffs and in the analysis and quality control of powders and dusts.
Specifications
  • Int.Ø×H:
    58×170 mm
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..