Assortment Boxes, PP
Assortment box, with hinged lid and 3×6 compartments, Anthracite, 240×195×40 mm
Catalog # SEMA1773
Supplier: SEMADENI
Jump to:
Product Details & Documents
These assortment boxes, made of PP, have a transparent stable hinged lid with lock and tamper-evident seals.
- Robust and strong design
- All models can be stacked together
- Lid is fixed on each tray so that the contents don't mix even if box is turned upside down
Specifications
- Colour:Anthracite
- Description:Assortment box, with hinged lid and 3×6 compartments
- Seq:04
- W×D×H:240×195×40 mm
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..