Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cryoboxes

Cryoboxes

without grid insert, -, Tissue culture tubes 15/50 ml, 148×148×128 mm
Catalog # NEOL2-7095
Supplier:   NEOLAB
Cryoboxes
Catalog # NEOL2-7095
Supplier:  NEOLAB
Cryoboxes
Cryoboxes

without grid insert, -, Tissue culture tubes 15/50 ml, 148×148×128 mm

Price
SFr. 16.20
Log in for availability
1 * 1 ST
Product Details & Documents

Cryo resistant cardboard.

  • Water repellent
  • Llid has a marking area on one of the sides

For storage of tissue culture tubes of 15 and 50 ml capacities for cryogenic deep freezers.

Specifications
  • Compartments:
    -
  • For:
    Tissue culture tubes 15/50 ml
  • W×D×H ext.:
    148×148×128 mm
  • Description:
    without grid insert
  • Seq:
    02
Accessories

Product Information

  • Catalog #
    Compartments
    For tubes
    Availability
    Price
  • Grid insert for cryo boxes
    Grid insert for cryo boxes
    NEOL2-7096
    5×5
    15 ml
    Log in for availability
    1 * 1 ST - SFr. 2.20

Product Information

  • Catalog #
    Compartments
    For tubes
    Availability
    Price
  • Grid insert for cryo boxes
    Grid insert for cryo boxes
    NEOL2-7098
    3×3
    50 ml
    Log in for availability
    1 * 1 ST - SFr. 2.60