Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Counting chambers, Sedgewick

Counting chambers, Sedgewick

Cell counter (plastic), 1 mm
Catalog # MARI0611400
Supplier:   MARIENFELD
Counting chambers, Sedgewick
Catalog # MARI0611400
Supplier:  MARIENFELD
Counting chambers, Sedgewick
Counting chambers, Sedgewick

Cell counter (plastic), 1 mm

Price
228.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

Counting chambers according to Sedgewick for particles and microorganisms in transparent liquids.

A cell is ruled in a grid subdividing 1,0 ml into 1000 μl.

Delivery information: Supplied with a cover glass.

Specifications
  • Depth of chamber:
    1 mm
  • Description:
    Cell counter (plastic)
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Cover glass
    Cover glass
    MARI0336000
    Log in for availability
    1 * 1 ST - 69.00 SFr.

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.