Orders LinkContactUsLinkComponent
Hoffmann clips
Hoffmann clips
20 mm
Catalog # MURRL141/02
Supplier:   MURRAY S
Hoffmann clips
Catalog # MURRL141/02
Supplier:  MURRAY S
Hoffmann clips
20 mm
Price
SFr. 22.30
Log in for availability
1 * 1 ST
Product Details & Documents
Bunsen screw clip, nickel plated brass fitted with protection plate and hinged bottom bar.
  • Spanning width = Width of gate
Specifications
  • Spanning width:
    20 mm
  • Seq:
    02