Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Test tubes without rim, medium wall

Test tubes without rim, medium wall

100 mm, 16 mm
Catalog # 212-0558
Supplier:   MURRAY S
Test tubes without rim, medium wall
Catalog # 212-0558
Supplier:  MURRAY S
Test tubes without rim, medium wall
Test tubes without rim, medium wall

100 mm, 16 mm

Price
SFr. 64.80
Log in for availability
1 * 100 ST
Product Details & Documents

Soda-lime glass, rimless.

  • Good chemical and physical properties
  • Medium-walled

Suitable for use in applications that do not involve high thermal stresses, especially for bacteriological applications.

Specifications
  • Ext.Ø:
    16 mm
  • Length:
    100 mm
  • Seq:
    09