Orders LinkContactUsLinkComponent
Test tubes without rim, medium wall
50 mm, 12 mm
  MURRG002/75
 :   MURRAY S
Test tubes without rim, medium wall
  MURRG002/75
 :  MURRAY S
Test tubes without rim, medium wall
Test tubes without rim, medium wall
50 mm, 12 mm
SFr. 44.90
1 * 100 ST
Soda-lime glass, rimless.
  • Good chemical and physical properties
  • Medium-walled
Suitable for use in applications that do not involve high thermal stresses, especially for bacteriological applications.
  • Ext.Ø:
    12 mm
  • Length:
    50 mm
  • Seq:
    03