Square HDPE Bottle
250 ml, Square bottle, HDPE
Catalog # SAMP8165A-250
Supplier: Sampling Systems
Jump to:
Product Details & Documents
Square HDPE bottles for use with a range of liquids and powders.
- Different sizes available
- Ribbed sides for easy handling
- Available from stock
- Fast delivery
This range of HDPE containers are perfect for use with a range of powders and liquids. They have a handy square space-saving shape and incorporate side ridges for easy handling when wet.
Specifications
- Capacity:250 ml
- Description:Square bottle
- Material:HDPE
- Seq:01