Orders LinkContactUsLinkComponent
Magnetic stirring bars, triangular
Magnetic stirring bars, triangular
25,32, 7,39 mm
Catalog # KART781
Supplier:   KARTELL
Magnetic stirring bars, triangular
Catalog # KART781
Supplier:  KARTELL
Magnetic stirring bars, triangular
Magnetic stirring bars, triangular
25,32, 7,39 mm
Price
SFr. 7.70
Log in for availability
1 * 1 ST
Product Details & Documents
PTFE coated, triangular magnetic stirring bars.
  • Scraping action ideal for dissolving solids and mixing sediments
  • Angled surfaces create high turbulence even at low speeds
  • Suitable for food stuff
Specifications
  • Ø:
    7,39 mm
  • Length:
    25,32
  • Seq:
    02