Orders LinkContactUsLinkComponent
Filter candles | Avantor
Filter candles
3, 13×25 mm
Katalognr. LENZ07206325
Anbieter:   Lenz Laborglas
Filter candles
Katalognr. LENZ07206325
Anbieter:  Lenz Laborglas
Filter candles
Filter candles
3, 13×25 mm
Preis
SFr. 56.40
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
These cylindrical filter plugs without tube are made of borosilicate glass 3,3.
Dokumente
Technische Daten
  • Ø×H:
    13×25 mm
  • Porosity:
    3
  • Seq:
    09