Filter candles
2, 13×25 mm
Catalog # LENZ07206225
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
These cylindrical filter plugs without tube are made of borosilicate glass 3,3.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Ø×H:13×25 mm
- Porosity:2
- Seq:08