Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Filter funnels

Filter funnels

50 ml, 35 mm, 1
Catalog # LENZ07195128
Supplier:   Lenz Laborglas
Filter funnels
Catalog # LENZ07195128
Supplier:  Lenz Laborglas
Filter funnels
Filter funnels

50 ml, 35 mm, 1

Price
SFr. 48.50
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

With integrated sintered glass filters disc.

Documents

Specifications
  • Plate Ø:
    35 mm
  • Porosity:
    1
  • Capacity:
    50 ml
  • Seq:
    01