Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Stirrer with paddles

Stirrer with paddles

29/32, 90* mm, 10 mm, 460 mm
Catalog # LENZ06308007
Supplier:   Lenz Laborglas
Stirrer with paddles
Catalog # LENZ06308007
Supplier:  Lenz Laborglas
Stirrer with paddles
Stirrer with paddles

29/32, 90* mm, 10 mm, 460 mm

Price
SFr. 142.00
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass shaft, PFTE paddles with ground and polished shaft.

Caution: * stirring Ø when blades extended

Specifications
  • NS:
    29/32
  • Seq:
    04
  • Shaft Ø:
    10 mm
  • Shaft length:
    460 mm
  • Stirrer Ø:
    90* mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.