Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tissue grinders, Potter type

Tissue grinders, Potter type

10 ml, 15×150 mm
Catalog # KART6110
Supplier:   KARTELL
Tissue grinders, Potter type
Catalog # KART6110
Supplier:  KARTELL
Tissue grinders, Potter type
Tissue grinders, Potter type

10 ml, 15×150 mm

Price
SFr. 129.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Int.Ø×H:
    15×150 mm
  • Capacity:
    10 ml
  • Seq:
    03