Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tissue grinders, Potter type

Tissue grinders, Potter type

2 ml, 8×120 mm
Catalog # KART6102
Supplier:   KARTELL
Tissue grinders, Potter type
Catalog # KART6102
Supplier:  KARTELL
Tissue grinders, Potter type
Tissue grinders, Potter type

2 ml, 8×120 mm

Price
SFr. 104.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Int.Ø×H:
    8×120 mm
  • Capacity:
    2 ml
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.