Orders LinkContactUsLinkComponent
Sodium nitrite ≥98%
Glass bottle for solids, 5.000 g
  A18668.0I
Sodium nitrite ≥98%
  A18668.0I
 :  7632-00-0
Sodium nitrite ≥98%
Sodium nitrite ≥98%
Glass bottle for solids, 5.000 g
 :  7632-00-0
278.00 SFr.
1 * 5.000 g
Purity: 98% CAS No.: 7632-00-0 Molecular Formula: NNaO2 Formula Weight: 69.00 Odour: Odourless Melting Point: 271° dec. UN No.: 1500 MDL No.: MFCD00011118
 :  Diazotizing salts
  •  :7632-00-0
  •  :NaNO₂
  •  :MFCD00011118
  •  :69 g/mol
  •  :14,8648
  •  :320 °C (1013 hPa)
  •  :271 °C (dec.)
  •  :2.17
  •  :Ambient
  •  :231-555-9
  •  :1500
  •  :5.1,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    5.000 g