Order Entry
Switzerland
Orders LinkContactUsLinkComponent
D-(-)-Ribose ≥98%

D-(-)-Ribose ≥98%

Glass bottle for solids, 10 g
Catalog # A17894.09
D-(-)-Ribose ≥98%
Catalog # A17894.09
CAS Number:  50-69-1
D-(-)-Ribose ≥98%
D-(-)-Ribose ≥98%

Glass bottle for solids, 10 g

CAS Number:  50-69-1
Price
SFr. 54.50
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 98% CAS No.: 50-69-1 Molecular Formula: C5H10O5 Formula Weight: 150.13 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 82-86° Beilstein Reference: 1723081 Merck Reference: 14,8204 Optical Rotation: -20° (c=10 in water, 28h) MDL No.: MFCD00135453

  • CAS Number:50-69-1
  • Formula:C₅H₁₀O₅
  • MDL Number:MFCD00135453
  • Molecular Weight:150.13 g/mol
  • Merck Index:14,8204
  • Boiling Pt:331 °C (1013 hPa)
  • Melting Pt:82…86 °C
  • Density:0.8 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-059-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications