Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bromothymol blue

Bromothymol blue

Glass bottle for solids, 250 g
Catalog # A17746.30
Bromothymol blue
Catalog # A17746.30
CAS Number:  76-59-5
Bromothymol blue
Bromothymol blue

Glass bottle for solids, 250 g

CAS Number:  76-59-5
Price
756.00 SFr.
Log in for availability
1 * 250 g
Product Details & Documents

CAS No.: 76-59-5 Molecular Formula: C27H28Br2O5S Formula Weight: 624.39 Melting Point: 200-202° Beilstein Reference: 373934 Merck Reference: 14,1445 MDL No.: MFCD00005872

Synonyms:  3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-59-5
  • Formula:C₂₇H₂₈Br₂O₅S
  • MDL Number:MFCD00005872
  • Molecular Weight:624.39 g/mol
  • Merck Index:14,1445
  • Melting Pt:200…202 °C
  • Storage Temperature:Ambient
  • EINECS:200-971-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications