Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bromothymol blue

Bromothymol blue

Glass bottle for solids, 10 g
Catalog # A17746.09
Bromothymol blue
Catalog # A17746.09
CAS Number:  76-59-5
Bromothymol blue
Bromothymol blue

Glass bottle for solids, 10 g

CAS Number:  76-59-5
Price
SFr. 66.90
Log in for availability
1 * 10 g
Product Details & Documents

CAS No.: 76-59-5 Molecular Formula: C27H28Br2O5S Formula Weight: 624.39 Melting Point: 200-202° Beilstein Reference: 373934 Merck Reference: 14,1445 MDL No.: MFCD00005872

Synonyms:  3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-59-5
  • Formula:C₂₇H₂₈Br₂O₅S
  • MDL Number:MFCD00005872
  • Molecular Weight:624.39 g/mol
  • Merck Index:14,1445
  • Melting Pt:200…202 °C
  • Storage Temperature:Ambient
  • EINECS:200-971-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.