Order Entry
Switzerland
Orders LinkContactUsLinkComponent
D-(-)-Fructose ≥99%

D-(-)-Fructose ≥99%

Glass bottle for solids, 1.000 g
Catalog # A17718.0B
D-(-)-Fructose ≥99%
Catalog # A17718.0B
CAS Number:  57-48-7
D-(-)-Fructose ≥99%
D-(-)-Fructose ≥99%

Glass bottle for solids, 1.000 g

CAS Number:  57-48-7
Price
SFr. 131.00
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 99% CAS No.: 57-48-7 Molecular Formula: C6H12O6 Formula Weight: 180.16 Odour: Odourless Melting Point: ca 105-110° Beilstein Reference: 1239004 Merck Reference: 14,4273 Optical Rotation: -89° (c=2 in water) MDL No.: MFCD00148910

Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,4273
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:∼ 105…110 °C
  • Density:1.59
  • Storage Temperature:Ambient
  • EINECS:200-333-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g

Specifications