Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Methyl red

Methyl red

Glass bottle for solids, 25 g
Catalog # A16690.14
Methyl red
Catalog # A16690.14
CAS Number:  493-52-7
Methyl red
Methyl red

Glass bottle for solids, 25 g

CAS Number:  493-52-7
Price
51.50 SFr.
Log in for availability
1 * 25 g
Product Details & Documents

CAS No.: 493-52-7 Molecular Formula: C15H15N3O2 Formula Weight: 269.30 Melting Point: 179-182° Beilstein Reference: 750102 Merck Reference: 14,6119 MDL No.: MFCD00002425

Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • Merck Index:14,6119
  • Melting Pt:179…182 °C
  • Density:1.31 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-776-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications