Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cyclohexanone ≥99%

Cyclohexanone ≥99%

Glass bottle, 2.500 mL
Catalog # A15607.0F
Flammable
Irritant
   Safer Human and Environmental Health
Cyclohexanone ≥99%
Catalog # A15607.0F
   Safer Human and Environmental Health
CAS Number:  108-94-1
Cyclohexanone ≥99%
Cyclohexanone ≥99%

Glass bottle, 2.500 mL

CAS Number:  108-94-1
Price
164.00 SFr.
Log in for availability
1 * 2.500 mL
Product Details & Documents

Purity: 99+% CAS No.: 108-94-1 Molecular Formula: C6H10O Formula Weight: 98.15 Flash Point: 47°(116°F) Refractive Index: 1.4507 Melting Point: -26° Boiling Point: 154-156° Beilstein Reference: 385735 Merck Reference: 14,2726 UN No.: 1915 MDL No.: MFCD00001625

Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:14,2726
  • Boiling Pt:154…156 °C
  • Melting Pt:–26 °C
  • Density:0.948
  • Flash Pt:47 °C (116 °F)
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • UN:1915
  • ADR:3,III
  • REACH:01-2119453616-35
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL
  • Cat. No.:
    A15607.0F
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.