Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Uridine ≥99%

Uridine ≥99%

Glass bottle for solids, 5 g
Catalog # A15227.06
Uridine ≥99%
Catalog # A15227.06
CAS Number:  58-96-8
Uridine ≥99%
Uridine ≥99%

Glass bottle for solids, 5 g

CAS Number:  58-96-8
Price
SFr. 75.60
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 99% CAS No.: 58-96-8 Molecular Formula: C9H12N2O6 Formula Weight: 244.20 Melting Point: 166-169° Beilstein Reference: 754904 Merck Reference: 14,9877 Optical Rotation: +9.5° (c=2 in water) MDL No.: MFCD00006526

Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • MDL Number:MFCD00006526
  • Molecular Weight:244.20 g/mol
  • Merck Index:14,9877
  • Melting Pt:166…169 °C
  • Storage Temperature:Ambient
  • EINECS:200-407-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.