Pentadecanoic acid ≥99%
Glass bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 1002-84-2 Molecular Formula: C15H30O2 Formula Weight: 242.40 Flash Point: 110°(230°F) Melting Point: 51-54° Boiling Point: 257°/100mm Beilstein Reference: 1773831 MDL No.: MFCD00002745
Synonyms: Pentadecylic acidn-Pentadecanoic acid
- CAS Number:1002-84-2
- Formula:C₁₅H₃₀O₂
- MDL Number:MFCD00002745
- Molecular Weight:242.4 g/mol
- Boiling Pt:257 °C (100 mmHg)
- Melting Pt:51…54 °C
- Density:0.8496 g/cm³ (70 °C)
- Flash Pt:110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:213-693-1
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



