Pentadecanoic acid ≥99%
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 1002-84-2 Molecular Formula: C15H30O2 Formula Weight: 242.40 Flash Point: 110°(230°F) Melting Point: 51-54° Boiling Point: 257°/100mm Beilstein Reference: 1773831 MDL No.: MFCD00002745
Synonyms: Pentadecylic acidn-Pentadecanoic acid
- CAS Number:1002-84-2
- Formula:C₁₅H₃₀O₂
- MDL Number:MFCD00002745
- Molecular Weight:242.4 g/mol
- Boiling Pt:257 °C (100 mmHg)
- Melting Pt:51…54 °C
- Density:0.8496 g/cm³ (70 °C)
- Flash Pt:110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:213-693-1
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



