1,3-Dihydroxyacetone dimer ≥97%
Glass bottle for solids, 1.000 g
Supplier: Thermo Fisher Scientific
1,3-Dihydroxyacetone dimer ≥97%
Catalog # A14189.0B
Supplier: Thermo Fisher Scientific
CAS Number: 62147-49-3
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 62147-49-3 Molecular Formula: C6H12O6 Formula Weight: 180.16 Storage Temperature: 2°C to 8°C Melting Point: ca 75-82° Beilstein Reference: 112910 Merck Reference: 14,3182 MDL No.: MFCD00051019
Synonyms: 2,5-Dihydroxy-1,4-dioxane-2,5-dimethanol
- CAS Number:62147-49-3
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00051019
- Molecular Weight:180.16 g/mol
- Merck Index:14,3182
- Melting Pt:∼ 75…82 °C
- EINECS:612-947-2
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1.000 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



