Caproic acid ≥98%
Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98+% CAS No.: 142-62-1 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 104°(219°F) Physical Form: Clear liquid Density: 0.929 Refractive Index: 1.4165 Appearance: Pale yellow Melting Point: -4° Boiling Point: 202-203° Beilstein Reference: 773837 Merck Reference: 14,1759 UN No.: 2829 MDL No.: MFCD00004421
Synonyms: Hexanoic acid
- CAS Number:142-62-1
- Formula:C₆H₁₂O₂
- MDL Number:MFCD00004421
- Molecular Weight:116.16 g/mol
- Merck Index:14,1759
- Boiling Pt:202…203 °C
- Melting Pt:–4 °C
- Density:0.929
- Flash Pt:104 °C (219 °F)
- Storage Temperature:Ambient
- EINECS:205-550-7
- UN:2829
- ADR:8,III
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL



