Celite® Standard Super-cel, filter aid
Glass bottle for solids, 2.500 g
Supplier: Thermo Fisher Scientific
Celite® Standard Super-cel, filter aid
Catalog # B21983.0E
Supplier: Thermo Fisher Scientific
CAS Number: 91053-39-3
Jump to:
Product Details & Documents
CAS No.: 61790-53-2 Odour: Odourless Merck Reference: 14,4973 MDL No.: MFCD00132803
- CAS Number:91053-39-3
- MDL Number:MFCD00132803
- Merck Index:14,4973
- Boiling Pt:2200 °C (1013 hPa)
- Melting Pt:1710 °C
- Density:2.3 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:319-127-6
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:2.500 g



