Behenic acid tech. 85%, Technical Grade
Glass bottle for solids, 5.000 g
Supplier: Thermo Fisher Scientific
Behenic acid tech. 85%, Technical Grade
Catalog # A12850.0I
Supplier: Thermo Fisher Scientific
CAS Number: 112-85-6
Jump to:
Product Details & Documents
Purity: 85% CAS No.: 112-85-6 Molecular Formula: CH3(CH2)20CO2H Formula Weight: 340.60 Melting Point: 74-78° Boiling Point: 306°/60mm Merck Reference: 14,1023 MDL No.: MFCD00002807
Synonyms: Docosanoic acid
- CAS Number:112-85-6
- Formula:C₂₂H₄₄O₂
- MDL Number:MFCD00002807
- Molecular Weight:340.59 g/mol
- Merck Index:14,1023
- Boiling Pt:306 °C (60 mmHg)
- Melting Pt:74…78 °C
- Density:0.8221 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-010-8
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5.000 g



