4-Methylmorpholine ≥99%
Glass bottle, 2.500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 109-02-4 Molecular Formula: C5H11NO Formula Weight: 101.15 Flash Point: 14°(57°F) Physical Form: Liquid Density: 0.920 Refractive Index: 1.4355 Appearance: Colourless Odour: Amine-like pH: 10.6 Melting Point: -65° Boiling Point: 116-118° Beilstein Reference: 102719 Merck Reference: 14,6277 UN No.: 2535 MDL No.: MFCD00006175
Synonyms: NMM, N-Methylmorpholine
- CAS Number:109-02-4
- Formula:C₅H₁₁NO
- MDL Number:MFCD00006175
- Molecular Weight:101.15 g/mol
- Merck Index:14,6277
- Boiling Pt:116…118 °C
- Melting Pt:–65 °C
- Density:0.920
- Flash Pt:14 °C (57 °F)
- Storage Temperature:Ambient
- EINECS:203-640-0
- UN:2535
- ADR:3,II
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2.500 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



