Caprinaldehyde 96%
Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 96% CAS No.: 112-31-2 Molecular Formula: C10H20O Formula Weight: 156.27 Flash Point: 85°(185°F) Refractive Index: 1.4280 Melting Point: -5° Boiling Point: 207-209° Beilstein Reference: 1362530 MDL No.: MFCD00007031
Synonyms: Decylaldehyde, Decanal, Decyl aldehyde
- CAS Number:112-31-2
- Formula:C₁₀H₂₀O
- MDL Number:MFCD00007031
- Molecular Weight:156.27 g/mol
- Boiling Pt:207…209 °C
- Melting Pt:–5 °C
- Density:0.828
- Flash Pt:85 °C (185 °F)
- Storage Temperature:Ambient
- EINECS:203-957-4
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL



