Hydroquinone ≥99%
Glass bottle for solids, 1.000 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 123-31-9 Molecular Formula: C6H6O2 Formula Weight: 110.11 Flash Point: 165°(329°F) Odour: Odourless pH: 3.75 Melting Point: 170-172° Boiling Point: 285-287° Beilstein Reference: 605970 Merck Reference: 14,4808 UN No.: 3077 MDL No.: MFCD00002339
Synonyms: 1,4-Dihydroxybenzene, Quinol
- CAS Number:123-31-9
- Formula:C₆H₄(OH)₂
- MDL Number:MFCD00002339
- Molecular Weight:110.11 g/mol
- Merck Index:14,4808
- Boiling Pt:285…287 °C
- Melting Pt:170…172 °C
- Density:1.320
- Flash Pt:165 °C (329 °F)
- Storage Temperature:Ambient
- EINECS:204-617-8
- UN:3077
- ADR:9,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1.000 g



