Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Calcium fluoride 97%, precipitated

Calcium fluoride 97%, precipitated

Glass bottle for solids, 250 g
Catalog # A10176.30
Irritant
Calcium fluoride 97%, precipitated
Catalog # A10176.30
CAS Number:  7789-75-5
Calcium fluoride 97%, precipitated
Calcium fluoride 97%, precipitated

Glass bottle for solids, 250 g

CAS Number:  7789-75-5
Price
SFr. 85.60
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 97% CAS No.: 7789-75-5 Molecular Formula: CaF2 Formula Weight: 78.08 Refractive Index: 1.434 Odour: Odourless Boiling Point: ca 2500° Merck Reference: 14,1667 MDL No.: MFCD00010907

  • CAS Number:7789-75-5
  • Formula:CaF₂
  • MDL Number:MFCD00010907
  • Molecular Weight:78.07 g/mol
  • Merck Index:14,1667
  • Boiling Pt:∼2500 °C
  • Melting Pt:1403 °C
  • Density:3.18
  • Storage Temperature:Ambient
  • EINECS:232-188-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.