Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Calcium fluoride 97%, precipitated

Glass bottle for solids, 5.000 g
Catalog # A10176.0I
Irritant
Calcium fluoride 97%, precipitated
Catalog # A10176.0I
CAS Number:  7789-75-5
Calcium fluoride 97%, precipitated
Calcium fluoride 97%, precipitated

Glass bottle for solids, 5.000 g

CAS Number:  7789-75-5
Price
SFr. 779.00
Log in for availability
1 * 5.000 g
Product Details & Documents

Purity: 97% CAS No.: 7789-75-5 Molecular Formula: CaF2 Formula Weight: 78.08 Refractive Index: 1.434 Odour: Odourless Boiling Point: ca 2500° Merck Reference: 14,1667 MDL No.: MFCD00010907

  • CAS Number:7789-75-5
  • Formula:CaF₂
  • MDL Number:MFCD00010907
  • Molecular Weight:78.07 g/mol
  • Merck Index:14,1667
  • Boiling Pt:∼2500 °C
  • Melting Pt:1403 °C
  • Density:3.18
  • Storage Temperature:Ambient
  • EINECS:232-188-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5.000 g

Specifications