Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tanks, rectangular, LDPE, LaboPlast®

Tanks, rectangular, LDPE, LaboPlast®

155 L, Seamless containers, rectangular shaped, 600×500×560 mm
Catalog # BURK3440-1556
Supplier:   BURKLE
Tanks, rectangular, LDPE, LaboPlast®
Catalog # BURK3440-1556
Supplier:  BURKLE
Tanks, rectangular, LDPE, LaboPlast®
Tanks, rectangular, LDPE, LaboPlast®

155 L, Seamless containers, rectangular shaped, 600×500×560 mm

Price
SFr. 600.00
Log in for availability
1 * 1 ST
Product Details & Documents

These seamless rectangular tanks feature optimum ratio of wall thickness to filling load.

  • Linear LDPE, transparent
  • Rupture-proof
Specifications
  • Capacity:
    155 L
  • Description:
    Seamless containers, rectangular shaped
  • Seq:
    13
  • W×D×H:
    600×500×560 mm
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Container thread ¾" for cylindrical containers
    Container thread ¾" for cylindrical containers
    BURK3440-0034
    Log in for availability
    1 * 1 ST - SFr. 202.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.