Stoppers, PTFE
White, 29/32
Catalog # BOHLH937-07
Supplier: Bohlender
Jump to:
Product Details & Documents
For closing ground joint parts. With ground joint and sealing rings on the outside; knurled or hexagonal grip. Compared with glass stoppers, they are easily removable and can be used without grease.
- PTFE
- Temperature resistance from −200 to + 250 °C
- Chemical resistance
- FDA conform
The stoppers can expand under heat which might lead to a breaking of the ground joint sockets.
Specifications
- NS:29/32
- Colour:White