ContactUsLinkComponent
Flask volumetric, capacity 10ml, wide neck, class A, amber graduated, poly stoppered, tolerance 0.04ml, height 90mm, NS-ground 10/19 , graduation color white, conformity certified. 1 * 1 items
Catalog # HIRS264016137
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 10ml, wide neck, class A, amber graduated, poly stoppered, tolerance 0.04ml, height 90mm, NS-ground 10/19 , graduation color white, conformity certified. 1 * 1 items
Catalog # HIRS264016137
Supplier:  HIRSCHMANN
Frequently Bought Together